Product Details

SNP ID
rs141998525
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5788384 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAGAAAAGTGAGGAACCCAGCTT[C/T]AGCAATGACTGAATTAGTGAGGATG
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
OR52N1 PubMed Links

Gene Details

Gene
OR52N1
Gene Name
olfactory receptor family 52 subfamily N member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001913.1 433 Missense Mutation AAA,GAA K145E NP_001001913.1
Gene
OR52N5
Gene Name
olfactory receptor family 52 subfamily N member 5
There are no transcripts associated with this gene.

View Full Product Details