Product Details

SNP ID
rs142030077
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59515023 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCCTGTTTTTGGTGTACATGAC[A/G]ACTCTAATGGGAAACTTCCTCATCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4D9 PubMed Links

Gene Details

Gene
OR4D9
Gene Name
olfactory receptor family 4 subfamily D member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004711.1 111 Silent Mutation ACA,ACG T37T NP_001004711.1

View Full Product Details