Product Details

SNP ID
rs142040344
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67405100 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCCCCAGGCCGGGCCACCCACC[C/T]GCAGACCTCATCCGCCAACGGGAGA
Phenotype
MIM: 176875 MIM: 603761 MIM: 610831
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPP1CA PubMed Links

Gene Details

Gene
PPP1CA
Gene Name
protein phosphatase 1 catalytic subunit alpha
There are no transcripts associated with this gene.

Gene
RAD9A
Gene Name
RAD9 checkpoint clamp component A
There are no transcripts associated with this gene.

Gene
TBC1D10C
Gene Name
TBC1 domain family member 10C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256508.1 484 Silent Mutation CCC,CCT P56P NP_001243437.1
NM_198517.3 484 Silent Mutation CCC,CCT P56P NP_940919.1
XM_006718538.3 484 Silent Mutation CCC,CCT P56P XP_006718601.1
XM_006718539.3 484 Silent Mutation CCC,CCT P56P XP_006718602.1
XM_006718540.3 484 Silent Mutation CCC,CCT P56P XP_006718603.1
XM_006718541.3 484 UTR 5 XP_006718604.1
XM_006718542.3 484 UTR 5 XP_006718605.1
XM_006718543.2 484 UTR 5 XP_006718606.1
XM_006718544.3 484 UTR 5 XP_006718607.1
XM_011545001.2 484 UTR 5 XP_011543303.1
XM_011545002.2 484 Silent Mutation CCC,CCT P56P XP_011543304.1

View Full Product Details