Product Details

SNP ID
rs142044984
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67487025 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACAGGCCACGTTCCACTACCGGA[C/T]GCTGCACAGTGACGACGAGGGCACC
Phenotype
MIM: 605555 MIM: 608794
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AIP PubMed Links

Gene Details

Gene
AIP
Gene Name
aryl hydrocarbon receptor interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302959.1 249 UTR 5 NP_001289888.1
NM_001302960.1 249 Missense Mutation ACG,ATG T40M NP_001289889.1
NM_003977.3 249 Missense Mutation ACG,ATG T40M NP_003968.3
Gene
MIR6752
Gene Name
microRNA 6752
There are no transcripts associated with this gene.

Gene
PITPNM1
Gene Name
phosphatidylinositol transfer protein membrane associated 1
There are no transcripts associated with this gene.

View Full Product Details