Product Details

SNP ID
rs142047066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:372196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCTGAGGACTCCAGTGAGAGCC[A/G]TGAAGAGGAGCAAGCGGTGAGCAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
B4GALNT4 PubMed Links

Gene Details

Gene
B4GALNT4
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178537.4 248 Missense Mutation CAT,CGT H80R NP_848632.2
XM_017017654.1 248 UTR 5 XP_016873143.1

View Full Product Details