Product Details

SNP ID
rs142116155
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56318550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAACAACTGTGGGACCCAAAATG[A/T]TAGTAAATTTTGTTGTGGATAAGAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR8K3 PubMed Links

Gene Details

Gene
OR8K3
Gene Name
olfactory receptor family 8 subfamily K member 3 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005202.1 244 Missense Mutation ATA,TTA I82L NP_001005202.1

View Full Product Details