Product Details

SNP ID
rs142164377
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64193128 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGGCCCTGAGCGTGGGTAACAT[C/T]GATGATGCCTTACAGTGCTACTCCG
Phenotype
MIM: 605063
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STIP1 PubMed Links

Gene Details

Gene
STIP1
Gene Name
stress induced phosphoprotein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282652.1 754 Silent Mutation ATC,ATT I67I NP_001269581.1
NM_001282653.1 754 Intron NP_001269582.1
NM_006819.2 754 Intron NP_006810.1

View Full Product Details