Product Details

SNP ID
rs142201801
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124440516 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACAAGCTCATTCACATAGGTGCT[A/G]GTGCAAGCACACTCAAGAAGGGGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR8B8 PubMed Links

Gene Details

Gene
OR8B8
Gene Name
olfactory receptor family 8 subfamily B member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012378.1 570 Silent Mutation ACC,ACT T190T NP_036510.1

View Full Product Details