Product Details

SNP ID
rs142245358
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72099214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGCTCCATTGAGAGCTTTGGTAG[G/T]GGGGCTGCTAGGGTCCAGCAGCAGC
Phenotype
MIM: 614717 MIM: 613510 MIM: 612414
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ANAPC15 PubMed Links

Gene Details

Gene
ANAPC15
Gene Name
anaphase promoting complex subunit 15
There are no transcripts associated with this gene.

Gene
LAMTOR1
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017907.2 248 Missense Mutation NP_060377.1
Gene
LRTOMT
Gene Name
leucine rich transmembrane and O-methyltransferase domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145307.4 248 Intron NP_001138779.1
NM_001145308.4 248 Intron NP_001138780.1
NM_001145309.3 248 Intron NP_001138781.1
NM_001145310.3 248 Intron NP_001138782.1
NM_001205138.3 248 Intron NP_001192067.1
NM_001271471.2 248 Intron NP_001258400.1
NM_001318803.1 248 Intron NP_001305732.1
NM_145309.5 248 Intron NP_660352.1
XM_006718473.3 248 Intron XP_006718536.1
XM_006718474.3 248 Intron XP_006718537.1
XM_011544847.2 248 Intron XP_011543149.1
XM_011544848.2 248 Intron XP_011543150.1
XM_017017356.1 248 Intron XP_016872845.1
XM_017017357.1 248 Intron XP_016872846.1
XM_017017358.1 248 Intron XP_016872847.1
XM_017017359.1 248 Intron XP_016872848.1
XM_017017360.1 248 Intron XP_016872849.1

View Full Product Details