Product Details

SNP ID
rs142345403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:7938592 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAACACAATCAGAATGTGTGTAAA[A/G]TCACTTTTCTTCGCCATAGTTTTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR10A3 PubMed Links

Gene Details

Gene
OR10A3
Gene Name
olfactory receptor family 10 subfamily A member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003745.1 Intron NP_001003745.1
Gene
OR10A6
Gene Name
olfactory receptor family 10 subfamily A member 6 (gene/pseudogene)
There are no transcripts associated with this gene.

View Full Product Details