Product Details

SNP ID
rs142432135
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6026989 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCTCAGGGCCTTCACTGCCGCC[C/G]CCTCTGCTTTGAATCTAAGCACAGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR56A1 PubMed Links

Gene Details

Gene
OR56A1
Gene Name
olfactory receptor family 56 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001917.2 753 Missense Mutation GCG,GGG A239G NP_001001917.2

View Full Product Details