Product Details
- SNP ID
-
rs142446933
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:129865101 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGATGTTGCGGCCCAACCCACTGAG[A/G]CTGTGGAGGGAAAGCAGGGGTGAGA
- Phenotype
-
MIM: 164013
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NFRKB
PubMed Links
Gene Details
- Gene
- NFRKB
- Gene Name
- nuclear factor related to kappaB binding protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001143835.1 |
3751 |
Silent Mutation |
AAC,AAT |
N1213N |
NP_001137307.1 |
NM_006165.3 |
3751 |
Silent Mutation |
AAC,AAT |
N1238N |
NP_006156.2 |
XM_005271567.3 |
3751 |
Silent Mutation |
AAC,AAT |
N1225N |
XP_005271624.1 |
XM_005271568.3 |
3751 |
Silent Mutation |
AAC,AAT |
N1225N |
XP_005271625.1 |
XM_011542850.2 |
3751 |
Silent Mutation |
AAC,AAT |
N1248N |
XP_011541152.1 |
XM_011542851.2 |
3751 |
Silent Mutation |
AAC,AAT |
N1238N |
XP_011541153.1 |
XM_011542852.2 |
3751 |
Silent Mutation |
AAC,AAT |
N1238N |
XP_011541154.1 |
XM_017017793.1 |
3751 |
Silent Mutation |
AAC,AAT |
N1255N |
XP_016873282.1 |
XM_017017794.1 |
3751 |
Silent Mutation |
AAC,AAT |
N1243N |
XP_016873283.1 |
XM_017017795.1 |
3751 |
Silent Mutation |
AAC,AAT |
N1236N |
XP_016873284.1 |
XM_017017796.1 |
3751 |
Silent Mutation |
AAC,AAT |
N1213N |
XP_016873285.1 |
XM_017017797.1 |
3751 |
Silent Mutation |
AAC,AAT |
N1166N |
XP_016873286.1 |
XM_017017798.1 |
3751 |
Silent Mutation |
AAC,AAT |
N1124N |
XP_016873287.1 |
XM_017017799.1 |
3751 |
Silent Mutation |
AAC,AAT |
N970N |
XP_016873288.1 |
XM_017017800.1 |
3751 |
Silent Mutation |
AAC,AAT |
N970N |
XP_016873289.1 |
XM_017017801.1 |
3751 |
Silent Mutation |
AAC,AAT |
N674N |
XP_016873290.1 |
- Gene
- TMEM45B
- Gene Name
- transmembrane protein 45B
There are no transcripts associated with this gene.
View Full Product Details