Product Details

SNP ID
rs142455569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57543884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCTCCAGATGGGCTGGGTCCAG[A/G]CTGTGTAGCTTCCGGACAGACCAGT
Phenotype
MIM: 613664 MIM: 603890
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMTNL1 PubMed Links

Gene Details

Gene
SMTNL1
Gene Name
smoothelin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105565.2 883 Missense Mutation GAC,GGC D294G NP_001099035.2
XM_005273819.3 883 Missense Mutation GAC,GGC D294G XP_005273876.1
Gene
UBE2L6
Gene Name
ubiquitin conjugating enzyme E2 L6
There are no transcripts associated with this gene.

View Full Product Details