Product Details

SNP ID
rs142476927
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57660028 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGTTTCTCTATCCAGGCCCCTCT[C/G]GTGTATCATTTTGGTTCCACCACTC
Phenotype
MIM: 608757 MIM: 614586
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CLP1 PubMed Links

Gene Details

Gene
CLP1
Gene Name
cleavage and polyadenylation factor I subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142597.1 691 Intron NP_001136069.1
NM_006831.2 691 Silent Mutation CTC,CTG L184L NP_006822.1
Gene
ZDHHC5
Gene Name
zinc finger DHHC-type containing 5
There are no transcripts associated with this gene.

View Full Product Details