Product Details

SNP ID
rs142509476
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67438753 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGCCCCTAGGTGGGAGGAGCCC[A/G]GGGCCATGGCGGGCCGGCTGTCAGA
Phenotype
MIM: 609849 MIM: 601577 MIM: 608939
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CORO1B PubMed Links
Additional Information
For this assay, SNP(s) [rs34026289] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CORO1B
Gene Name
coronin 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018070.2 1373 Missense Mutation CCG,CTG P421L NP_001018080.1
NM_020441.2 1373 Missense Mutation CCG,CTG P421L NP_065174.1
Gene
PTPRCAP
Gene Name
protein tyrosine phosphatase, receptor type C associated protein
There are no transcripts associated with this gene.

Gene
RPS6KB2
Gene Name
ribosomal protein S6 kinase B2
There are no transcripts associated with this gene.

View Full Product Details