Product Details
- SNP ID
-
rs142536582
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:46645388 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGCTTGGTGAAAAGATTTGCACTC[A/G]TTCATCATCTTCTCCAACGGGTTCA
- Phenotype
-
MIM: 615088
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ATG13
PubMed Links
Gene Details
- Gene
- ATG13
- Gene Name
- autophagy related 13
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001142673.2 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
NP_001136145.1 |
NM_001205119.1 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
NP_001192048.1 |
NM_001205120.1 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
NP_001192049.1 |
NM_001205121.1 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
NP_001192050.1 |
NM_001205122.1 |
825 |
UTR 5 |
|
|
NP_001192051.1 |
NM_014741.4 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
NP_055556.2 |
XM_005253262.2 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_005253319.1 |
XM_005253263.2 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_005253320.1 |
XM_005253265.2 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_005253322.1 |
XM_005253266.2 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_005253323.1 |
XM_005253268.2 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_005253325.1 |
XM_006718394.2 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_006718457.1 |
XM_006718395.2 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_006718458.1 |
XM_006718396.2 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_006718459.1 |
XM_011520493.1 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_011518795.1 |
XM_011520496.2 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_011518798.1 |
XM_011520499.2 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_011518801.1 |
XM_011520500.1 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_011518802.1 |
XM_011520501.1 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_011518803.1 |
XM_011520502.1 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_011518804.1 |
XM_011520503.1 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_011518805.1 |
XM_017018599.1 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_016874088.1 |
XM_017018600.1 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_016874089.1 |
XM_017018601.1 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_016874090.1 |
XM_017018602.1 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_016874091.1 |
XM_017018603.1 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_016874092.1 |
XM_017018604.1 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_016874093.1 |
XM_017018605.1 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_016874094.1 |
XM_017018606.1 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_016874095.1 |
XM_017018607.1 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_016874096.1 |
XM_017018608.1 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_016874097.1 |
XM_017018609.1 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_016874098.1 |
XM_017018610.1 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_016874099.1 |
XM_017018611.1 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_016874100.1 |
XM_017018612.1 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_016874101.1 |
XM_017018613.1 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_016874102.1 |
XM_017018614.1 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_016874103.1 |
XM_017018615.1 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_016874104.1 |
XM_017018616.1 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_016874105.1 |
XM_017018617.1 |
825 |
Missense Mutation |
CAT,CGT |
H40R |
XP_016874106.1 |
XM_017018618.1 |
825 |
UTR 5 |
|
|
XP_016874107.1 |
XM_017018619.1 |
825 |
UTR 5 |
|
|
XP_016874108.1 |
XM_017018620.1 |
825 |
UTR 5 |
|
|
XP_016874109.1 |
XM_017018621.1 |
825 |
UTR 5 |
|
|
XP_016874110.1 |
XM_017018622.1 |
825 |
UTR 5 |
|
|
XP_016874111.1 |
View Full Product Details