Product Details

SNP ID
rs142686850
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:109424213 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGGCGAGGGCCTTTTGCAAACG[A/G]TGGTACTGTCCTGATCGTCTAGCCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C11orf87 PubMed Links

Gene Details

Gene
C11orf87
Gene Name
chromosome 11 open reading frame 87
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207645.3 1087 Missense Mutation ATG,GTG M194V NP_997528.2
XM_011542817.2 1087 Missense Mutation ATG,GTG M341V XP_011541119.1
XM_011542818.2 1087 Missense Mutation ATG,GTG M194V XP_011541120.1

View Full Product Details