Product Details

SNP ID
rs142738268
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56743103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGGATTCTCAGGATAGCCAGGA[A/G]GATGTTGACATAGGAAATCAGGATA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6128 PubMed Links

Gene Details

Gene
MIR6128
Gene Name
microRNA 6128
There are no transcripts associated with this gene.

Gene
OR9G4
Gene Name
olfactory receptor family 9 subfamily G member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005284.1 709 Missense Mutation CTC,TTC L237F NP_001005284.1

View Full Product Details