Product Details

SNP ID
rs142792520
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:129376080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGCTGAGCTCGCCCGGCCAGCT[C/G]AAAGCAGCCAGGCGGCGCTACAAGA
Phenotype
MIM: 604823
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BARX2 PubMed Links

Gene Details

Gene
BARX2
Gene Name
BARX homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003658.4 95 Silent Mutation CTC,CTG L15L NP_003649.2
XM_011543043.1 95 Intron XP_011541345.1
XM_011543044.2 95 Intron XP_011541346.1

View Full Product Details