Product Details

SNP ID
rs142792552
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55820115 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCATGTTCCTTGGAACTGACGTG[C/T]TCTGCTTTAAAGTTATGTTTTCATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5D18 PubMed Links

Gene Details

Gene
OR5D18
Gene Name
olfactory receptor family 5 subfamily D member 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001952.1 486 Silent Mutation TGC,TGT C162C NP_001001952.1
Gene
OR5L1
Gene Name
olfactory receptor family 5 subfamily L member 1 (gene/pseudogene)
There are no transcripts associated with this gene.

Gene
OR5L2
Gene Name
olfactory receptor family 5 subfamily L member 2
There are no transcripts associated with this gene.

View Full Product Details