Product Details

SNP ID
rs142808709
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:870430 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAAAACACTCACTTCTTGTACTC[G/T]AAGAAGTGCTCTGAGGCCTGGCTGT
Phenotype
MIM: 615692 MIM: 602644
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CHID1 PubMed Links

Gene Details

Gene
CHID1
Gene Name
chitinase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142674.1 1247 Silent Mutation NP_001136146.1
NM_001142675.1 1247 Silent Mutation NP_001136147.1
NM_001142676.1 1247 Silent Mutation NP_001136148.1
NM_001142677.1 1247 Silent Mutation NP_001136149.1
NM_023947.3 1247 Silent Mutation NP_076436.3
XM_005253072.2 1247 Silent Mutation XP_005253129.1
XM_005253073.4 1247 Silent Mutation XP_005253130.1
XM_006718282.2 1247 Silent Mutation XP_006718345.1
XM_011520299.1 1247 Silent Mutation XP_011518601.1
XM_011520300.1 1247 Silent Mutation XP_011518602.1
XM_011520301.2 1247 Intron XP_011518603.1
XM_011520302.2 1247 Intron XP_011518604.1
XM_017018166.1 1247 Silent Mutation XP_016873655.1
XM_017018167.1 1247 Intron XP_016873656.1
XM_017018168.1 1247 Intron XP_016873657.1
Gene
TSPAN4
Gene Name
tetraspanin 4
There are no transcripts associated with this gene.

View Full Product Details