Product Details

SNP ID
rs142889509
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64227067 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGATGACTTCCTTGTCTTCCTGCGC[C/T]GCTCCCGGCAGGTGGCTGAGGCCCC
Phenotype
MIM: 604189 MIM: 607901 MIM: 610470 MIM: 601398
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC4 PubMed Links

Gene Details

Gene
DNAJC4
Gene Name
DnaJ heat shock protein family (Hsp40) member C4
There are no transcripts associated with this gene.

Gene
FERMT3
Gene Name
fermitin family member 3
There are no transcripts associated with this gene.

Gene
NUDT22
Gene Name
nudix hydrolase 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128612.2 473 Missense Mutation CGC,TGC R139C NP_001122084.1
NM_001128613.2 473 Missense Mutation CGC,TGC R139C NP_001122085.1
NM_001271831.1 473 Missense Mutation CGC,TGC R139C NP_001258760.1
NM_032344.3 473 Missense Mutation CGC,TGC R139C NP_115720.1
Gene
TRPT1
Gene Name
tRNA phosphotransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033678.3 473 Intron NP_001028850.2
NM_001160389.1 473 Intron NP_001153861.1
NM_001160390.1 473 Intron NP_001153862.1
NM_001160392.1 473 Intron NP_001153864.1
NM_001160393.1 473 Intron NP_001153865.1
NM_031472.3 473 Intron NP_113660.1
XM_005274345.4 473 Intron XP_005274402.1
XM_005274346.4 473 Intron XP_005274403.1
XM_005274347.4 473 Intron XP_005274404.1
XM_006718706.3 473 Intron XP_006718769.1
XM_006718707.3 473 Intron XP_006718770.1
XM_017018400.1 473 Intron XP_016873889.1
XM_017018401.1 473 Intron XP_016873890.1
Gene
VEGFB
Gene Name
vascular endothelial growth factor B
There are no transcripts associated with this gene.

View Full Product Details