Product Details

SNP ID
rs142893022
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67304776 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTGCCCACTGCCCTGCCAGGCA[A/G]AGCTTTGCGGTGCTCCGTGGGGCTG
Phenotype
MIM: 615126 MIM: 606780
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD13D PubMed Links

Gene Details

Gene
ANKRD13D
Gene Name
ankyrin repeat domain 13D
There are no transcripts associated with this gene.

Gene
SSH3
Gene Name
slingshot protein phosphatase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017857.3 286 Silent Mutation CAA,CAG Q36Q NP_060327.3
XM_017017942.1 286 UTR 5 XP_016873431.1

View Full Product Details