Product Details

SNP ID
rs142984715
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63817986 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCTGAGATCCGGGCACCCTCGGC[C/T]GACACAGCTCGCTCGCACATCTTGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf84 PubMed Links

Gene Details

Gene
C11orf84
Gene Name
chromosome 11 open reading frame 84
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138471.2 622 Silent Mutation GCC,GCT A103A NP_612480.1
XM_005273782.2 622 Silent Mutation GCC,GCT A103A XP_005273839.1
XM_005273783.2 622 Silent Mutation GCC,GCT A103A XP_005273840.1
XM_006718437.1 622 Silent Mutation GCC,GCT A103A XP_006718500.1
XM_011544770.1 622 Silent Mutation GCC,GCT A103A XP_011543072.1
XM_011544771.1 622 Silent Mutation GCC,GCT A103A XP_011543073.1
XM_011544772.1 622 Silent Mutation GCC,GCT A103A XP_011543074.1

View Full Product Details