Product Details

SNP ID
rs143087131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55651129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACATTTGTTACTCTTCAGTCACA[C/G]CTCCCAAGATGATTGTTGACCTGTT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR4S2 PubMed Links

Gene Details

Gene
OR4S2
Gene Name
olfactory receptor family 4 subfamily S member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004059.2 226 Missense Mutation CCT,GCT P76A NP_001004059.2

View Full Product Details