Product Details

SNP ID
rs143152014
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:112060024 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTAATGATGTTGTTTCTTTAGCA[A/G]CCAAAGCAAGAGAGGGTAAACTACA
Phenotype
MIM: 608770
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DLAT PubMed Links

Gene Details

Gene
DLAT
Gene Name
dihydrolipoamide S-acetyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001931.4 2295 Missense Mutation ACC,GCC T546A NP_001922.2
XM_011542647.2 2295 Missense Mutation ACC,GCC T510A XP_011540949.1
Gene
PIH1D2
Gene Name
PIH1 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001082619.1 2295 Intron NP_001076088.1
NM_138789.3 2295 Intron NP_620144.1
XM_011542593.2 2295 Intron XP_011540895.2
XM_017017201.1 2295 Intron XP_016872690.1
XM_017017202.1 2295 Intron XP_016872691.1
XM_017017203.1 2295 Intron XP_016872692.1
XM_017017204.1 2295 Intron XP_016872693.1
XM_017017205.1 2295 Intron XP_016872694.1

View Full Product Details