Product Details

SNP ID
rs143154193
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64361476 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCTCTTGCCCTCCCAGAAAGAGC[A/G]GACCTTCTCCTTCTGTGGCACCATC
Phenotype
MIM: 611205 MIM: 603606
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC88B PubMed Links

Gene Details

Gene
CCDC88B
Gene Name
coiled-coil domain containing 88B
There are no transcripts associated with this gene.

Gene
MIR1237
Gene Name
microRNA 1237
There are no transcripts associated with this gene.

Gene
RPS6KA4
Gene Name
ribosomal protein S6 kinase A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006944.1 641 Missense Mutation CAG,CGG Q193R NP_001006945.1
NM_001300802.1 641 Missense Mutation CAG,CGG Q193R NP_001287731.1
NM_001318361.1 641 Missense Mutation CAG,CGG Q130R NP_001305290.1
NM_003942.2 641 Missense Mutation CAG,CGG Q193R NP_003933.1
XM_005274380.1 641 Missense Mutation CAG,CGG Q193R XP_005274437.1
XM_017018527.1 641 UTR 5 XP_016874016.1

View Full Product Details