Product Details

SNP ID
rs143213326
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:69699473 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTACAGCTGCCGCTGTTTGGCA[C/G]TGCTCAGGGAGACCGGGAGGCGGTG
Phenotype
MIM: 603891
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FGF19 PubMed Links

Gene Details

Gene
FGF19
Gene Name
fibroblast growth factor 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005117.2 903 Missense Mutation ACT,AGT T147S NP_005108.1

View Full Product Details