Product Details

SNP ID
rs143217870
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:13962945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCCCCTGAAGCTGAGCCGGACTC[A/C]GGCACTGCTGGCCCTGGCGCTGCCC
Phenotype
MIM: 604989
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SPON1 PubMed Links

Gene Details

Gene
SPON1
Gene Name
spondin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006108.3 309 Missense Mutation CAG,CCG Q14P NP_006099.2

View Full Product Details