Product Details

SNP ID
rs143230449
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:48245108 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTTGGCTAACATACACAACATGA[A/C]TGAATTCATTTTTCTGGTACTTTCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR4X2 PubMed Links

Gene Details

Gene
OR4X2
Gene Name
olfactory receptor family 4 subfamily X member 2 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004727.1 5 Missense Mutation AAT,ACT N2T NP_001004727.1

View Full Product Details