Product Details

SNP ID
rs143481066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47273892 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTTGGTGCCCTGATGCTTCTCTG[A/T]GATAAACTGATGAATTGGAACCATG
Phenotype
MIM: 603584 MIM: 602423
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC101928943 PubMed Links

Gene Details

Gene
LOC101928943
Gene Name
uncharacterized LOC101928943
There are no transcripts associated with this gene.

Gene
MADD
Gene Name
MAP kinase activating death domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135943.1 193 UTR 5 NP_001129415.1
NM_001135944.1 193 UTR 5 NP_001129416.1
NM_003682.3 193 UTR 5 NP_003673.3
NM_130470.2 193 UTR 5 NP_569826.2
NM_130471.2 193 UTR 5 NP_569827.2
NM_130472.2 193 UTR 5 NP_569828.2
NM_130473.2 193 UTR 5 NP_569829.2
NM_130474.2 193 UTR 5 NP_569830.2
NM_130475.2 193 UTR 5 NP_569831.1
NM_130476.2 193 UTR 5 NP_569832.2
XM_005253189.2 193 UTR 5 XP_005253246.1
XM_005253196.2 193 UTR 5 XP_005253253.1
XM_005253199.2 193 UTR 5 XP_005253256.1
XM_005253200.2 193 UTR 5 XP_005253257.1
XM_005253201.2 193 UTR 5 XP_005253258.1
XM_005253203.2 193 UTR 5 XP_005253260.1
XM_005253204.2 193 UTR 5 XP_005253261.1
XM_005253205.1 193 UTR 5 XP_005253262.1
XM_017018478.1 193 UTR 5 XP_016873967.1
XM_017018479.1 193 UTR 5 XP_016873968.1
XM_017018480.1 193 UTR 5 XP_016873969.1
XM_017018481.1 193 UTR 5 XP_016873970.1
XM_017018482.1 193 UTR 5 XP_016873971.1
XM_017018483.1 193 UTR 5 XP_016873972.1
XM_017018484.1 193 UTR 5 XP_016873973.1
XM_017018485.1 193 UTR 5 XP_016873974.1
XM_017018486.1 193 UTR 5 XP_016873975.1
XM_017018487.1 193 UTR 5 XP_016873976.1
XM_017018488.1 193 UTR 5 XP_016873977.1
XM_017018489.1 193 UTR 5 XP_016873978.1
XM_017018490.1 193 UTR 5 XP_016873979.1
XM_017018491.1 193 UTR 5 XP_016873980.1
XM_017018492.1 193 UTR 5 XP_016873981.1
XM_017018493.1 193 UTR 5 XP_016873982.1
XM_017018494.1 193 UTR 5 XP_016873983.1
XM_017018495.1 193 UTR 5 XP_016873984.1
XM_017018496.1 193 UTR 5 XP_016873985.1
XM_017018497.1 193 UTR 5 XP_016873986.1
XM_017018498.1 193 UTR 5 XP_016873987.1
XM_017018499.1 193 UTR 5 XP_016873988.1
XM_017018500.1 193 UTR 5 XP_016873989.1
XM_017018501.1 193 UTR 5 XP_016873990.1
XM_017018502.1 193 UTR 5 XP_016873991.1
XM_017018503.1 193 UTR 5 XP_016873992.1
XM_017018504.1 193 UTR 5 XP_016873993.1
XM_017018505.1 193 UTR 5 XP_016873994.1
XM_017018506.1 193 UTR 5 XP_016873995.1
XM_017018507.1 193 UTR 5 XP_016873996.1
XM_017018508.1 193 UTR 5 XP_016873997.1
XM_017018509.1 193 UTR 5 XP_016873998.1
XM_017018510.1 193 UTR 5 XP_016873999.1
Gene
NR1H3
Gene Name
nuclear receptor subfamily 1 group H member 3
There are no transcripts associated with this gene.

View Full Product Details