Product Details

SNP ID
rs143495423
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66526159 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCTTTGCTTTGGCCGGTACGGG[A/C]GGGAGGACAACACCCTCATCATGAC
Phenotype
MIM: 209901
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BBS1 PubMed Links

Gene Details

Gene
BBS1
Gene Name
Bardet-Biedl syndrome 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024649.4 1159 Silent Mutation AGG,CGG R383R NP_078925.3
Gene
ZDHHC24
Gene Name
zinc finger DHHC-type containing 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207340.1 1159 Intron NP_997223.1
XM_005273874.3 1159 Intron XP_005273931.1
XM_011544893.2 1159 Intron XP_011543195.1
XM_011544894.1 1159 Intron XP_011543196.1

View Full Product Details