Product Details

SNP ID
rs143574894
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:130228280 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGATATAAGCATTAGAAGTCATCA[C/T]TTTGTACAAGGAAGAAAATGAATAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZBTB44 PubMed Links

Gene Details

Gene
ZBTB44
Gene Name
zinc finger and BTB domain containing 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301098.1 7757 UTR 3 NP_001288027.1
NM_001301099.1 7757 UTR 3 NP_001288028.1
NM_014155.4 7757 UTR 3 NP_054874.3
XM_005271521.4 7757 UTR 3 XP_005271578.1
XM_005271522.4 7757 UTR 3 XP_005271579.1
XM_006718825.3 7757 UTR 3 XP_006718888.1
XM_006718826.3 7757 UTR 3 XP_006718889.1
XM_011542789.2 7757 Intron XP_011541091.1
XM_017017623.1 7757 UTR 3 XP_016873112.1
XM_017017624.1 7757 UTR 3 XP_016873113.1

View Full Product Details