Product Details

SNP ID
rs143598977
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4881994 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCTCATGTCTGGATCTCCATTC[C/T]AGTCTGCTGTCTCTACACCATTGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR51T1 PubMed Links

Gene Details

Gene
OR51T1
Gene Name
olfactory receptor family 51 subfamily T member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004759.1 176 Missense Mutation CCA,CTA P59L NP_001004759.1

View Full Product Details