Product Details

SNP ID
rs143600911
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:17720305 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGACGCCGCGCCCCCTGGCGCC[G/T]CAGCCGCCTTCTATGCGCCGGGCCC
Phenotype
MIM: 159970
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MYOD1 PubMed Links

Gene Details

Gene
MYOD1
Gene Name
myogenic differentiation 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002478.4 743 Missense Mutation GCA,TCA A175S NP_002469.2

View Full Product Details