Product Details

SNP ID
rs143648646
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58402801 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACCAGGAGAGCTATAAAGATATT[A/G]AAGCTCACAACATAAATAAGAACAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5B3 PubMed Links

Gene Details

Gene
OR5B3
Gene Name
olfactory receptor family 5 subfamily B member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005469.1 609 Silent Mutation TTC,TTT F203F NP_001005469.1

View Full Product Details