Product Details

SNP ID
rs143767955
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:130230886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTGAACATCTTTGTTTACTTCT[A/G]AAGTACATCACACCTATTTGGTAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZBTB44 PubMed Links

Gene Details

Gene
ZBTB44
Gene Name
zinc finger and BTB domain containing 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301098.1 5151 UTR 3 NP_001288027.1
NM_001301099.1 5151 UTR 3 NP_001288028.1
NM_014155.4 5151 UTR 3 NP_054874.3
XM_005271521.4 5151 UTR 3 XP_005271578.1
XM_005271522.4 5151 UTR 3 XP_005271579.1
XM_006718825.3 5151 UTR 3 XP_006718888.1
XM_006718826.3 5151 UTR 3 XP_006718889.1
XM_011542789.2 5151 Intron XP_011541091.1
XM_017017623.1 5151 UTR 3 XP_016873112.1
XM_017017624.1 5151 UTR 3 XP_016873113.1

View Full Product Details