Product Details

SNP ID
rs143791296
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6768056 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGTATTTTCCCAGGACCCTCCTC[A/G]AGGCCCGCATGACCTCCTTATTCCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2AG2 PubMed Links

Gene Details

Gene
OR2AG2
Gene Name
olfactory receptor family 2 subfamily AG member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004490.1 Intron NP_001004490.1

View Full Product Details