Product Details

SNP ID
rs143866203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67608659 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAAGACATCGGGTTTGAGGGGCC[A/G]TGGAGGCGCTGGCTTCCCCACTGGC
Phenotype
MIM: 161015
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C11orf72 PubMed Links

Gene Details

Gene
C11orf72
Gene Name
chromosome 11 open reading frame 72
There are no transcripts associated with this gene.

Gene
DOC2GP
Gene Name
double C2 domain gamma pseudogene
There are no transcripts associated with this gene.

Gene
NDUFV1
Gene Name
NADH:ubiquinone oxidoreductase core subunit V1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166102.1 416 Missense Mutation CAT,CGT H79R NP_001159574.1
NM_007103.3 416 Missense Mutation CAT,CGT H88R NP_009034.2

View Full Product Details