Product Details

SNP ID
rs143882371
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:8615938 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTCACAGGATGGTGGGAATGTGG[A/G]CTGCACAAAGGAAAGATGACCTAGA
Phenotype
MIM: 612000
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM66 PubMed Links

Gene Details

Gene
TRIM66
Gene Name
tripartite motif containing 66
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014818.1 6706 UTR 3 NP_055633.1
XM_006718397.2 6706 UTR 3 XP_006718460.1
XM_006718398.3 6706 UTR 3 XP_006718461.1
XM_011520504.1 6706 UTR 3 XP_011518806.1
XM_011520507.2 6706 UTR 3 XP_011518809.1
XM_011520508.1 6706 UTR 3 XP_011518810.1
XM_011520509.1 6706 UTR 3 XP_011518811.1
XM_011520510.1 6706 UTR 3 XP_011518812.1
XM_011520511.1 6706 UTR 3 XP_011518813.1
XM_011520512.1 6706 UTR 3 XP_011518814.1
XM_011520513.1 6706 UTR 3 XP_011518815.1
XM_011520514.2 6706 UTR 3 XP_011518816.1
XM_011520515.1 6706 UTR 3 XP_011518817.1
XM_011520516.1 6706 UTR 3 XP_011518818.1
XM_011520517.2 6706 UTR 3 XP_011518819.1
XM_011520518.1 6706 UTR 3 XP_011518820.1
XM_011520519.1 6706 UTR 3 XP_011518821.1
XM_011520522.2 6706 UTR 3 XP_011518824.1
XM_011520523.2 6706 UTR 3 XP_011518825.1
XM_011520524.1 6706 Intron XP_011518826.1
XM_011520525.1 6706 Intron XP_011518827.1
XM_011520526.2 6706 UTR 3 XP_011518828.1
XM_011520527.1 6706 UTR 3 XP_011518829.1
XM_017018629.1 6706 UTR 3 XP_016874118.1
XM_017018630.1 6706 UTR 3 XP_016874119.1
XM_017018631.1 6706 UTR 3 XP_016874120.1

View Full Product Details