Product Details
- SNP ID
-
rs143910613
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:62605924 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGCATTCTTCGTGGTTCCCACCAG[C/G]AGCTCAGAGCCAAGCCCTTCAGCAA
- Phenotype
-
MIM: 606374
MIM: 603947
MIM: 180721
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
B3GAT3
PubMed Links
Gene Details
- Gene
- B3GAT3
- Gene Name
- beta-1,3-glucuronyltransferase 3
There are no transcripts associated with this gene.
- Gene
- EML3
- Gene Name
- echinoderm microtubule associated protein like 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001300793.1 |
2396 |
Silent Mutation |
CTC,CTG |
L572L |
NP_001287722.1 |
NM_001300794.1 |
2396 |
Silent Mutation |
CTC,CTG |
L571L |
NP_001287723.1 |
NM_153265.2 |
2396 |
Silent Mutation |
CTC,CTG |
L571L |
NP_694997.2 |
XM_005273878.3 |
2396 |
Silent Mutation |
CTC,CTG |
L572L |
XP_005273935.1 |
XM_006718489.3 |
2396 |
Silent Mutation |
CTC,CTG |
L622L |
XP_006718552.1 |
XM_006718490.3 |
2396 |
Silent Mutation |
CTC,CTG |
L622L |
XP_006718553.1 |
XM_006718491.3 |
2396 |
Silent Mutation |
CTC,CTG |
L621L |
XP_006718554.1 |
XM_011544896.2 |
2396 |
Silent Mutation |
CTC,CTG |
L621L |
XP_011543198.1 |
XM_011544897.2 |
2396 |
Silent Mutation |
CTC,CTG |
L570L |
XP_011543199.1 |
XM_017017480.1 |
2396 |
Silent Mutation |
CTC,CTG |
L455L |
XP_016872969.1 |
XM_017017481.1 |
2396 |
Silent Mutation |
CTC,CTG |
L455L |
XP_016872970.1 |
XM_017017482.1 |
2396 |
Silent Mutation |
CTC,CTG |
L455L |
XP_016872971.1 |
XM_017017483.1 |
2396 |
Silent Mutation |
CTC,CTG |
L354L |
XP_016872972.1 |
- Gene
- MTA2
- Gene Name
- metastasis associated 1 family member 2
There are no transcripts associated with this gene.
- Gene
- ROM1
- Gene Name
- retinal outer segment membrane protein 1
There are no transcripts associated with this gene.
View Full Product Details