Product Details

SNP ID
rs143977407
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57659578 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTGGAGGCATCTCAGTCAGTTCA[G/T]TTGGAGTTGTTGACTGGCATGGCAG
Phenotype
MIM: 608757 MIM: 609725 MIM: 614586
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CLP1 PubMed Links

Gene Details

Gene
CLP1
Gene Name
cleavage and polyadenylation factor I subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142597.1 241 Missense Mutation CAG,CAT Q34H NP_001136069.1
NM_006831.2 241 Missense Mutation CAG,CAT Q34H NP_006822.1
Gene
YPEL4
Gene Name
yippee like 4
There are no transcripts associated with this gene.

Gene
ZDHHC5
Gene Name
zinc finger DHHC-type containing 5
There are no transcripts associated with this gene.

View Full Product Details