Product Details

SNP ID
rs144006817
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:54603733 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCATGCATCCATTGAATAGGATG[G/A]CCTTCTTTCCATAGAGTGAATGTGT
Phenotype
MIM: 614273
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
OR4C46 PubMed Links

Gene Details

Gene
OR4C46
Gene Name
olfactory receptor family 4 subfamily C member 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004703.1 Intron NP_001004703.1

View Full Product Details