Product Details

SNP ID
rs144026711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:69295831 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGGCTCAGCCAGGAGGCAGAAGA[C/T]GTGGACGTGTCCCGGGCCAGGAGGG
Phenotype
MIM: 605625
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYEOV PubMed Links

Gene Details

Gene
MYEOV
Gene Name
myeloma overexpressed
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293291.1 679 Silent Mutation GAC,GAT D127D NP_001280220.1
NM_001293294.1 679 Silent Mutation GAC,GAT D69D NP_001280223.1
NM_001293296.1 679 Silent Mutation GAC,GAT D69D NP_001280225.1
NM_001300923.1 679 Silent Mutation GAC,GAT D69D NP_001287852.1
NM_001300924.1 679 Silent Mutation GAC,GAT D69D NP_001287853.1
NM_138768.3 679 Silent Mutation GAC,GAT D127D NP_620123.2

View Full Product Details