Product Details

SNP ID
rs144081005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6768503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTATGTCCTATAGCAATCAGGGAT[C/G]CCAGGATCCAGGATGTGGCCACCAT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR2AG2 PubMed Links

Gene Details

Gene
OR2AG2
Gene Name
olfactory receptor family 2 subfamily AG member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004490.1 Intron NP_001004490.1

View Full Product Details