Product Details

SNP ID
rs144134779
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62752679 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACACTGCTCCTCCAGATGGGAAA[G/T]CTCTCTCAGGACCCTGAGGCTGTTC
Phenotype
MIM: 602013
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
POLR2G PubMed Links

Gene Details

Gene
POLR2G
Gene Name
RNA polymerase II subunit G
There are no transcripts associated with this gene.

Gene
ZBTB3
Gene Name
zinc finger and BTB domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024784.3 1080 Intron NP_079060.1
XM_011545256.2 1080 Missense Mutation GAT,GCT D329A XP_011543558.1

View Full Product Details