Product Details

SNP ID
rs144145921
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:12683369 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGGAGGTTATACATTGTGGATG[A/G]GTGTTGCCTTGAGGGTTGCTTTGAG
Phenotype
MIM: 189967
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TEAD1 PubMed Links

Gene Details

Gene
TEAD1
Gene Name
TEA domain transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021961.5 Intron NP_068780.2

View Full Product Details