Product Details

SNP ID
rs144270884
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:36490261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACAGTTTGTACCCGGGTTTGCC[A/G]GTGTAGAATCCAGGGCTATGAATCA
Phenotype
MIM: 602355
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRAF6 PubMed Links

Gene Details

Gene
TRAF6
Gene Name
TNF receptor associated factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004620.3 1434 Silent Mutation ACC,ACT T382T NP_004611.1
NM_145803.2 1434 Silent Mutation ACC,ACT T382T NP_665802.1
XM_017018220.1 1434 Intron XP_016873709.1

View Full Product Details