Product Details

SNP ID
rs144279571
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72577387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGAAGTGTCCCTGGAGGGGATCA[C/T]TCAGCATCAAGGCTGCAGCAGCCAT
Phenotype
MIM: 602658
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01537 PubMed Links

Gene Details

Gene
LINC01537
Gene Name
long intergenic non-protein coding RNA 1537
There are no transcripts associated with this gene.

Gene
PDE2A
Gene Name
phosphodiesterase 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143839.3 3302 Silent Mutation GAA,GAG E934E NP_001137311.1
NM_001146209.2 3302 Silent Mutation GAA,GAG E932E NP_001139681.1
NM_001243784.1 3302 Silent Mutation GAA,GAG E920E NP_001230713.1
NM_002599.4 3302 Silent Mutation GAA,GAG E941E NP_002590.1
XM_005274040.2 3302 Silent Mutation GAA,GAG E940E XP_005274097.1
XM_011545096.2 3302 Silent Mutation GAA,GAG E620E XP_011543398.1
XM_017017910.1 3302 Silent Mutation GAA,GAG E620E XP_016873399.1

View Full Product Details