Product Details

SNP ID
rs144429777
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:82733626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCTCATCGTCCTCGAAACAGCAG[C/G]CCTTGTCCAGGGCTCCGAAGGCGCC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM181B PubMed Links

Gene Details

Gene
FAM181B
Gene Name
family with sequence similarity 181 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175885.3 239 Missense Mutation GCC,GGC A35G NP_787081.2

View Full Product Details